N,N,4-Trimethylpyridin-2-amine 95%
Supplier: Apollo Scientific
OR942385-1G OR942385-250MG
APOSOR942385-1GEA 574 EUR
APOSOR942385-1G APOSOR942385-250MG
N,N,4-Trimethylpyridin-2-amine 95%
N,N,4-Trimethylpyridin-2-amine
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015432
CAS Number: 20173-72-2