6-Methoxy-imidazo[1,2-b]pyridazine 95%
Supplier: Apollo Scientific
OR942662-1G OR942662-250MG
APOSOR942662-1GEA 734 EUR
APOSOR942662-1G APOSOR942662-250MG
6-Methoxy-imidazo[1,2-b]pyridazine 95%
6-Methoxy-imidazo[1,2-b]pyridazine
Formula: C₇H₇N₃O
MW: 149.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12913581
CAS Number: 17240-33-4