5,6-Dihydro-7H-pyrrolo[1,2-c]imidazol-7-one ≥98%
Supplier: Apollo Scientific
OR943014-100MG OR943014-250MG OR943014-50MG
APOSOR943014-100MGEA 909 EUR
APOSOR943014-100MG APOSOR943014-250MG APOSOR943014-50MG
5,6-Dihydro-7H-pyrrolo[1,2-c]imidazol-7-one ≥98%
5,6-Dihydro-7H-pyrrolo[1,2-c]imidazol-7-one
Formula: C₆H₆N₂O
MW: 122.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17013182
CAS Number: 426219-43-4