2-Chloro-4-methoxy-6-methylpyrimidine 98%
Supplier: Apollo Scientific
OR943132-1G OR943132-250MG
APOSOR943132-1GEA 95.7 EUR
APOSOR943132-1G APOSOR943132-250MG
2-Chloro-4-methoxy-6-methylpyrimidine 98%
2-Chloro-4-methoxy-6-methylpyrimidine
Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689881
CAS Number: 22536-64-7