5-Methylpyrazolo[1,5-a]pyrimidin-7-ol 95%
Supplier: Apollo Scientific
OR943542-1G OR943542-250MG
APOSOR943542-1GEA 754 EUR
APOSOR943542-1G APOSOR943542-250MG
5-Methylpyrazolo[1,5-a]pyrimidin-7-ol 95%
5-Methylpyrazolo[1,5-a]pyrimidin-7-ol
Formula: C₇H₇N₃O
MW: 149.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00104730
CAS Number: 16082-26-1