(5-Methylimidazo[1,2-a]pyridin-2-yl)methanol 95%
Supplier: Apollo Scientific
OR943667-1G OR943667-5G
APOSOR943667-1GEA 319 EUR
APOSOR943667-1G APOSOR943667-5G
(5-Methylimidazo[1,2-a]pyridin-2-yl)methanol 95%
(5-Methylimidazo[1,2-a]pyridin-2-yl)methanol
Formula: C₉H₁₀N₂O
MW: 162.188 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863392
CAS Number: 872363-02-5