(5-Methylimidazo[1,2-a]pyridin-3-yl)methanol 95%
Supplier: Apollo Scientific
OR943823-1G OR943823-5G
APOSOR943823-1GEA 63.8 EUR
APOSOR943823-1G APOSOR943823-5G
(5-Methylimidazo[1,2-a]pyridin-3-yl)methanol 95%
(5-Methylimidazo[1,2-a]pyridin-3-yl)methanol
Formula: C₉H₁₀N₂O
MW: 162.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864459
CAS Number: 178488-39-6