1-(2,6-Dichloropyridin-4-yl)ethanone 95%
Supplier: Apollo Scientific
OR943953-1G
APOSOR943953-1GEA 82.8 EUR
APOSOR943953-1G
1-(2,6-Dichloropyridin-4-yl)ethanone 95%
1-(2,6-Dichloropyridin-4-yl)ethanone
Formula: C₇H₅Cl₂NO
MW: 190.027 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847255
CAS Number: 185319-20-4