[3-(Thien-2-yl)-1H-pyrazol-1-yl]acetic acid
Supplier: Apollo Scientific
Warning
OR9446-100MG OR9446-250MG
APOSOR9446-100MGEA 40.8 EUR
APOSOR9446-100MG APOSOR9446-250MG
[3-(Thien-2-yl)-1H-pyrazol-1-yl]acetic acid
[3-(Thien-2-yl)-1H-pyrazol-1-yl]acetic acid
Formula: C₉H₈N₂O₂S
MW: 208.237 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665861
CAS Number: 959582-96-8