2-Chloro-7-iodo-5-methyl-5H-pyrrolo[3,2-d]pyrimidine ≥98%
Supplier: Apollo Scientific
OR946102-100MG OR946102-1G
APOSOR946102-100MGEA 287 EUR
APOSOR946102-100MG APOSOR946102-1G
2-Chloro-7-iodo-5-methyl-5H-pyrrolo[3,2-d]pyrimidine ≥98%
2-Chloro-7-iodo-5-methyl-5H-pyrrolo[3,2-d]pyrimidine
Formula: C₇H₅ClIN₃
MW: 293.492 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19689071
CAS Number: 1152475-62-1