4-Chloro-1,2-benzoxazol-3-amine 98%
Supplier: Apollo Scientific
OR946341-1G OR946341-5G
APOSOR946341-1GEA 219 EUR
APOSOR946341-1G APOSOR946341-5G
4-Chloro-1,2-benzoxazol-3-amine 98%
4-Chloro-1,2-benzoxazol-3-amine
Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11186205
CAS Number: 868271-15-2