5-Chloro-1,2-benzoxazol-3-amine 97%
Supplier: Apollo Scientific
OR946342-1G OR946342-5G
APOSOR946342-1GEA 208 EUR
APOSOR946342-1G APOSOR946342-5G
5-Chloro-1,2-benzoxazol-3-amine 97%
5-Chloro-1,2-benzoxazol-3-amine
Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999136
CAS Number: 73498-24-5