5-Oxo-1-phenylpyrrolidine-3-carbonitrile 96%

Supplier: Apollo Scientific

OR946384-10G OR946384-1G OR946384-5G
APOSOR946384-10GEA 754 EUR
APOSOR946384-10G APOSOR946384-1G APOSOR946384-5G
5-Oxo-1-phenylpyrrolidine-3-carbonitrile 96%
5-Oxo-1-phenylpyrrolidine-3-carbonitrile
Formula: C₁₁H₁₀N₂O
MW: 186.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10004117
CAS Number: 120937-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR