1,2-Dimethyl-3-propoxybenzene 98%
Supplier: Apollo Scientific
OR946391-1G OR946391-5G
APOSOR946391-1GEA 183 EUR
APOSOR946391-1G APOSOR946391-5G
1,2-Dimethyl-3-propoxybenzene 98%
1,2-Dimethyl-3-propoxybenzene
Formula: C₁₁H₁₆O
MW: 164.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26386335
CAS Number: 833460-92-7