2-(2-Methoxyethoxy)benzaldehyde 98%
Supplier: Apollo Scientific
OR946445-1G OR946445-250MG
APOSOR946445-1GEA 364 EUR
APOSOR946445-1G APOSOR946445-250MG
2-(2-Methoxyethoxy)benzaldehyde 98%
2-(2-Methoxyethoxy)benzaldehyde
Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09036531
CAS Number: 92637-96-2