2-(3-Methoxyphenyl)-N-methylacetamide 98%

Supplier: Apollo Scientific

OR946456-1G OR946456-5G
APOSOR946456-1GEA 203 EUR
APOSOR946456-1G APOSOR946456-5G
2-(3-Methoxyphenyl)-N-methylacetamide 98%
2-(3-Methoxyphenyl)-N-methylacetamide
Formula: C₁₀H₁₃NO₂
MW: 179.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16557464
CAS Number: 53102-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR