N-Methyl-1,4-dioxaspiro[4.5]decan-8-amine 95%
Supplier: Apollo Scientific
OR947243-1G
OR947243-250MG
APOSOR947243-1GEA
754
EUR
APOSOR947243-1G
APOSOR947243-250MG
N-Methyl-1,4-dioxaspiro[4.5]decan-8-amine 95%
N-Methyl-1,4-dioxaspiro[4.5]decan-8-amine
Formula:
C₉H₁₇NO₂ MW: 171.237 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09733329 CAS Number: 166398-41-0 |