N-Propylacrylamide 95%

Supplier: Apollo Scientific

OR947270-1G OR947270-5G
APOSOR947270-1GEA 42.2 EUR
APOSOR947270-1G APOSOR947270-5G
N-Propylacrylamide 95%
N-Propylacrylamide
Formula: C₆H₁₁NO
MW: 113.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00080629
CAS Number: 25999-13-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR