1,3-Diazaspiro[4.6]undecane-2,4-dione 95%
Supplier: Apollo Scientific
OR948189-1G OR948189-5G
APOSOR948189-1GEA 458 EUR
APOSOR948189-1G APOSOR948189-5G
1,3-Diazaspiro[4.6]undecane-2,4-dione 95%
1,3-Diazaspiro[4.6]undecane-2,4-dione
Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022405
CAS Number: 707-16-4