8-Methoxychroman-4-one 95%
Supplier: Apollo Scientific
OR948291-1G
APOSOR948291-1GEA
108
EUR
APOSOR948291-1G
8-Methoxychroman-4-one 95%
8-Methoxychroman-4-one
Formula:
C₁₀H₁₀O₃ MW: 178.185 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00094977 CAS Number: 20351-79-5 |