Methyl 2,2-dimethyl-3-oxocyclobutane-1-carboxylate
Supplier: Apollo Scientific
OR949100-1G OR949100-250MG OR949100-500MG
APOSOR949100-1GEA 1490 EUR
APOSOR949100-1G APOSOR949100-250MG APOSOR949100-500MG
Methyl 2,2-dimethyl-3-oxocyclobutane-1-carboxylate
Methyl 2,2-dimethyl-3-oxocyclobutane-1-carboxylate
Formula: C₈H₁₂O₃
MW: 156.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21332873
CAS Number: 78685-51-5