2-Azaspiro[4.5]decane-7-carboxylic acid
Supplier: Apollo Scientific
OR949228-1G OR949228-250MG OR949228-500MG
APOSOR949228-1GEA 1110 EUR
APOSOR949228-1G APOSOR949228-250MG APOSOR949228-500MG
2-Azaspiro[4.5]decane-7-carboxylic acid
2-Azaspiro[4.5]decane-7-carboxylic acid
Formula: C₁₀H₁₇NO₂
MW: 183.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22566122
CAS Number: 1363382-89-1