6-Azaspiro[2.5]octan-4-one hydrochloride
Supplier: Apollo Scientific
OR949325-1G OR949325-250MG OR949325-500MG
APOSOR949325-1GEA 1710 EUR
APOSOR949325-1G APOSOR949325-250MG APOSOR949325-500MG
6-Azaspiro[2.5]octan-4-one hydrochloride
6-Azaspiro[2.5]octan-4-one hydrochloride
Formula: C₇H₁₁NO·ClH
MW: 161.629 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23105969
CAS Number: 1408076-12-9