Methyl 3-hydroxy-2,2-dimethylcyclobutane-1-carboxylate
Supplier: Apollo Scientific
OR949363-1G OR949363-250MG OR949363-500MG
APOSOR949363-1GEA 766 EUR
APOSOR949363-1G APOSOR949363-250MG APOSOR949363-500MG
Methyl 3-hydroxy-2,2-dimethylcyclobutane-1-carboxylate
Methyl 3-hydroxy-2,2-dimethylcyclobutane-1-carboxylate
Formula: C₈H₁₄O₃
MW: 158.195 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23106161
CAS Number: 527751-16-2