2,4-Dimethyl-3-nitrobenzonitrile 95%
Supplier: Apollo Scientific
OR949900-1G OR949900-250MG
APOSOR949900-1GEA 313 EUR
APOSOR949900-1G APOSOR949900-250MG
2,4-Dimethyl-3-nitrobenzonitrile 95%
2,4-Dimethyl-3-nitrobenzonitrile
Formula: C₉H₈N₂O₂
MW: 176.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08693500
CAS Number: 39053-46-8