2-Bromo-1-(2,2-dimethyl-4H-benzo[d][1,3]dioxin-6-yl)ethanone ≥98%
Supplier: Apollo Scientific
OR950408-1G OR950408-250MG
APOSOR950408-1GEA 246 EUR
APOSOR950408-1G APOSOR950408-250MG
2-Bromo-1-(2,2-dimethyl-4H-benzo[d][1,3]dioxin-6-yl)ethanone ≥98%
2-Bromo-1-(2,2-dimethyl-4H-benzo[d][1,3]dioxin-6-yl)ethanone
Formula: C₁₂H₁₃BrO₃
MW: 285.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26385854
CAS Number: 102293-80-1