(2,6-Dichloro-3-methylphenyl)methanol 95%
Supplier: Apollo Scientific
OR950707-1G OR950707-250MG
APOSOR950707-1GEA 134 EUR
APOSOR950707-1G APOSOR950707-250MG
(2,6-Dichloro-3-methylphenyl)methanol 95%
(2,6-Dichloro-3-methylphenyl)methanol
Formula: C₈H₈Cl₂O
Storage Temperature: Ambient
MDL Number: MFCD09999718
CAS Number: 1378814-84-6