[(Adamantan-1-yl)methyl](methyl)amine 95%
Supplier: Apollo Scientific
Warning
OR951105-1G OR951105-5G
APOSOR951105-1GEA 376 EUR
APOSOR951105-1G APOSOR951105-5G
[(Adamantan-1-yl)methyl](methyl)amine 95%
[(Adamantan-1-yl)methyl](methyl)amine
Formula: C₁₂H₂₁N
MW: 179.302 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07658364
CAS Number: 153461-22-4