2-Allyl-4-methoxyphenol 95%
Supplier: Apollo Scientific
OR951404-1G OR951404-250MG
APOSOR951404-1GEA 435 EUR
APOSOR951404-1G APOSOR951404-250MG
2-Allyl-4-methoxyphenol 95%
2-Allyl-4-methoxyphenol
Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00996134
CAS Number: 584-82-7