4-Chloro-3-methylquinoline 95%
Supplier: Apollo Scientific
OR951626-1G OR951626-250MG
APOSOR951626-1GEA 89.9 EUR
APOSOR951626-1G APOSOR951626-250MG
4-Chloro-3-methylquinoline 95%
4-Chloro-3-methylquinoline
Formula: C₁₀H₈ClN
MW: 177.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09264040
CAS Number: 63136-60-7