3-Amino-5-ethoxyisoxazole 95%
Supplier: Apollo Scientific
OR951656-100MG OR951656-1G OR951656-250MG
APOSOR951656-100MGEA 145 EUR
APOSOR951656-100MG APOSOR951656-1G APOSOR951656-250MG
3-Amino-5-ethoxyisoxazole 95%
3-Amino-5-ethoxyisoxazole
Formula: C₅H₈N₂O₂
MW: 128.129 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14584669
CAS Number: 32326-26-4