7H-Pyrrolo[2,3-d]pyrimidine-4-carbonitrile 95%
Supplier: Apollo Scientific
OR951668-100MG OR951668-1G OR951668-250MG
APOSOR951668-100MGEA 161 EUR
APOSOR951668-100MG APOSOR951668-1G APOSOR951668-250MG
7H-Pyrrolo[2,3-d]pyrimidine-4-carbonitrile 95%
7H-Pyrrolo[2,3-d]pyrimidine-4-carbonitrile
Formula: C₇H₄N₄
MW: 144.133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17015948
CAS Number: 1005206-16-5