2-[4-(Methylthio)phenoxy]ethylamine 95%
Supplier: Apollo Scientific
OR951874-1G OR951874-250MG
APOSOR951874-1GEA 364 EUR
APOSOR951874-1G APOSOR951874-250MG
2-[4-(Methylthio)phenoxy]ethylamine 95%
2-[4-(Methylthio)phenoxy]ethylamine
Formula: C₉H₁₃NOS
MW: 183.271 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13370835
CAS Number: 1203188-22-0