5-Nitroquinolin-2(1H)-one 95%
Supplier: Apollo Scientific
OR951924-1G OR951924-250MG
APOSOR951924-1GEA 647 EUR
APOSOR951924-1G APOSOR951924-250MG
5-Nitroquinolin-2(1H)-one 95%
5-Nitroquinolin-2(1H)-one
Formula: C₉H₆N₂O₃
MW: 190.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11110268
CAS Number: 6938-27-8