6-(Propan-2-yl)-2,3-dihydropyridazin-3-one 95%
Supplier: Apollo Scientific
Synonyms: 6-Isopropylpyridazin-3(2H)-one
OR951953-1G OR951953-250MG
APOSOR951953-1GEA 951 EUR
APOSOR951953-1G APOSOR951953-250MG
6-(Propan-2-yl)-2,3-dihydropyridazin-3-one 95%
6-(Propan-2-yl)-2,3-dihydropyridazin-3-one
Formula: C₇H₁₀N₂O
MW: 138.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11616599
CAS Number: 570416-36-3