Thieno[3,2-d]pyrimidin-4-ol 95%
Supplier: Apollo Scientific
OR952006-100G OR952006-25G OR952006-5G
APOSOR952006-100GEA 719 EUR
APOSOR952006-100G APOSOR952006-25G APOSOR952006-5G
Thieno[3,2-d]pyrimidin-4-ol 95%
Thieno[3,2-d]pyrimidin-4-ol
Formula: C₆H₄N₂OS
MW: 152.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040452
CAS Number: 14080-55-8