4-Chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Apollo Scientific
OR952021-1G OR952021-250MG
APOSOR952021-1GEA 696 EUR
APOSOR952021-1G APOSOR952021-250MG
4-Chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine 95%
4-Chloro-3-nitro-1H-pyrrolo[2,3-b]pyridine
Formula: C₇H₄ClN₃O₂
MW: 197.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09880131
CAS Number: 918519-53-6