2-Chloro-1-(4-ethylphenyl)-ethanone 95%
Supplier: Apollo Scientific
OR952123-1G OR952123-5G
APOSOR952123-1GEA 502 EUR
APOSOR952123-1G APOSOR952123-5G
2-Chloro-1-(4-ethylphenyl)-ethanone 95%
2-Chloro-1-(4-ethylphenyl)-ethanone
Formula: C₁₀H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD03964194
CAS Number: 50690-09-0