2-(1H-Pyrazol-3-yl)acetonitrile 95%
Supplier: Apollo Scientific
OR952219-1G OR952219-250MG
APOSOR952219-1GEA 806 EUR
APOSOR952219-1G APOSOR952219-250MG
2-(1H-Pyrazol-3-yl)acetonitrile 95%
2-(1H-Pyrazol-3-yl)acetonitrile
Formula: C₅H₅N₃
MW: 107.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08436151
CAS Number: 135237-01-3