4,7-Dimethoxy-1-indanone 95%
Supplier: Apollo Scientific
Synonyms: 4,7-Dimethoxy-2,3-dihydro-1H-inden-1-one
OR952308-5G
APOSOR952308-5GEA 133 EUR
APOSOR952308-5G
4,7-Dimethoxy-1-indanone 95%
4,7-Dimethoxy-1-indanone
Formula: C₁₁H₁₂O₃
MW: 192.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00757244
CAS Number: 52428-09-8