Ethyl 1-methylimidazole-4-carboxylate 98%
Supplier: Apollo Scientific
OR952460-1G OR952460-5G
APOSOR952460-1GEA 37.7 EUR
APOSOR952460-1G APOSOR952460-5G
Ethyl 1-methylimidazole-4-carboxylate 98%
Ethyl 1-methylimidazole-4-carboxylate
Formula: C₇H₁₀N₂O₂
MW: 154.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223488
CAS Number: 41507-56-6