6-Pyridin-4-yl-2H-pyridazin-3-one 97%
Supplier: Apollo Scientific
Synonyms: 6-(Pyridin-4-yl)pyridazin-3(2H)-one
OR952502-1G OR952502-250MG
APOSOR952502-1GEA 314 EUR
APOSOR952502-1G APOSOR952502-250MG
6-Pyridin-4-yl-2H-pyridazin-3-one 97%
6-Pyridin-4-yl-2H-pyridazin-3-one
Formula: C₉H₇N₃O
MW: 173.171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00205154
CAS Number: 78157-26-3