7-Chloro-2-quinoxalinone 95%

Supplier: Apollo Scientific

OR952590-1G OR952590-250MG OR952590-5G
APOSOR952590-1GEA 270 EUR
APOSOR952590-1G APOSOR952590-250MG APOSOR952590-5G
7-Chloro-2-quinoxalinone 95%
7-Chloro-2-quinoxalinone
Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800423
CAS Number: 59489-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR