8-Benzyl-2,8-diazaspiro[4.5]decan-3-one
Supplier: Apollo Scientific
OR952704-5G
APOSOR952704-5GEA 79.9 EUR
APOSOR952704-5G
8-Benzyl-2,8-diazaspiro[4.5]decan-3-one
8-Benzyl-2,8-diazaspiro[4.5]decan-3-one
Formula: C₁₅H₂₀N₂O
MW: 244.332 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15147047
CAS Number: 154495-69-9