Methyl 6-methylpyridazine-3-carboxylate 95%
Supplier: Apollo Scientific
OR952867-1G OR952867-250MG
APOSOR952867-1GEA 142 EUR
APOSOR952867-1G APOSOR952867-250MG
Methyl 6-methylpyridazine-3-carboxylate 95%
Methyl 6-methylpyridazine-3-carboxylate
Formula: C₇H₈N₂O₂
MW: 152.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13174778
CAS Number: 106584-51-4