Chroman-4-one oxime 95%
Supplier: Apollo Scientific
OR952980-1G OR952980-250MG
APOSOR952980-1GEA 754 EUR
APOSOR952980-1G APOSOR952980-250MG
Chroman-4-one oxime 95%
Chroman-4-one oxime
Formula: C₉H₉NO₂
MW: 163.173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463254
CAS Number: 24541-01-3