5-Aminopyrimidine-4,6-diol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-4,6-dihydroxypyrimidine
OR953017-1G OR953017-25G OR953017-5G
APOSOR953017-1GEA 53.8 EUR
APOSOR953017-1G APOSOR953017-25G APOSOR953017-5G
5-Aminopyrimidine-4,6-diol 97%
5-Aminopyrimidine-4,6-diol
Formula: C₄H₅N₃O₂
MW: 127.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00239457
CAS Number: 69340-97-2