Methyl 5-(methylthio)nicotinate 95%
Supplier: Apollo Scientific
OR953103-1G OR953103-250MG
APOSOR953103-1GEA 435 EUR
APOSOR953103-1G APOSOR953103-250MG
Methyl 5-(methylthio)nicotinate 95%
Methyl 5-(methylthio)nicotinate
Formula: C₈H₉NO₂S
MW: 183.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18070989
CAS Number: 74470-43-2