(S)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%
Supplier: Apollo Scientific
Synonyms: (S)-Rasagiline
OR953162-1G OR953162-250MG
APOSOR953162-1GEA 1720 EUR
APOSOR953162-1G APOSOR953162-250MG
(S)-N-(2-Propynyl)-2,3-dihydroinden-1-amine 95%
(S)-N-(2-Propynyl)-2,3-dihydroinden-1-amine
Formula: C₁₂H₁₃N
MW: 171.238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029306
CAS Number: 185517-74-2