2-(Methylthio)-6-phenylpyrimidin-4-ol
Supplier: Apollo Scientific
Warning
Synonyms: 2-(Methysulphanyl)-6-phenylpyrimidin-4-ol, 2-(Methylsulphanyl)-6-phenylpyrimidin-4(1H)-one, 2-(Methylsulphanyl)-6-phenylpyrimidin-4(3H)-one
OR9532-1G OR9532-500MG
APOSOR9532-1GEA 219 EUR
APOSOR9532-1G APOSOR9532-500MG
2-(Methylthio)-6-phenylpyrimidin-4-ol
2-(Methylthio)-6-phenylpyrimidin-4-ol
Formula: C₁₁H₁₀N₂OS
MW: 218.275 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00124285
CAS Number: 56035-29-1